Z677922
Finntip® Flex pipette tips
Finntip Flex 1000, sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
1 mL
volume range:
100-1000 μL
sterility:
sterile
feature:
barrier: no
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Finn 94060713
Doporučené produkty
material
blue
colorless tip
sterility
sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Finn 94060713
maximum volume
1 mL
volume range
100-1000 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)