Hlavní dokumenty
Z675520
Captor® pen holder
allows one-handed uncapping and capping of marker
About This Item
UNSPSC Code:
41103032
NACRES:
NB.35
material
black polycarbonate
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art F13373-0000
W × D × H
25 mm (1 in.) × 25 mm (1 in.) × 51 mm (2 in.)
General description
Affix Captor Pen Holder wherever a marker pen is regularly needed. The fixed location allows one-handed uncapping and capping of marker.
This clever accessory reduces mishandling with samples and writing instruments when marking and placing items into frequently used equipment such as an autoclave or freezer.
Other possible mounting locations include the incubator, centrifuge, storeroom or next to the lab bench. The adhesive backed, press-fit Dual Lock re-closable fastener affixes to almost any clean, flat surface. The penholder works with most pen caps that have a pocket clip.
Legal Information
Captor is a registered trademark of SP Scienceware, a division of SP
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Zákazníci si také prohlíželi
Slide 1 of 1
1 of 1
Advanced PAP Pen 2 mm tip width
Z672548
Advanced PAP Pen