Z671703
Rare earth extra power stir bar set, Cylindrical
14 stir bars total
About This Item
Doporučené produkty
material
PTFE-coated
cylindrical
packaging
pack of 14 ea
manufacturer/tradename
Cowie 001.3036.RE
General description
set contents:-
2 each of 8x3, 12x4.5, 20x6, 25x6, 30x6 & 40x8
1 of 50x8 & 60x10
2 each of 8x3, 12x4.5, 20x6, 25x6, 30x6 & 40x8
1 of 50x8 & 60x10