Key Documents
Z556254
Sigma-Aldrich
Extractor body for Aldrich® Soxhlet extractor
capacity 200 mL, top joint: ST/NS 55/50, bottom joint: ST/NS 24/40
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
joint
bottom joint: ST/NS24/40
top joint: ST/NS55/50
capacity
200 mL