Z553662
Aldrich® Liebig condenser with SafetyBarb® hose connectors
Overall L 200 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Doporučené produkty
joint
joint ST/NS24/40
Overall L
200 mm
General description
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany