Key Documents
Z552704
Sigma-Aldrich
Aldrich® Thermometer Adapter
straight, male joint: ST/NS 19/22, GL 14 thread
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
male joint: ST/NS19/22
fitting
GL 14 thread
General description
Has PTFE faced silicone washer and open-top cap.