Key Documents
Z544221
Molymod® Atoms and Bonds
Halogen, no. of holes, 4, diam. 23 mm, angle, 109.50
About This Item
UNSPSC Code:
60104505
NACRES:
NB.17
feature
angle 109.50
no. of holes 4
manufacturer/tradename
Molymod
diam.
23 mm
color
green
Packaging
1PAK = 10EA
10PAK = 10 x 1PAK or 100EA
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.