Hlavní dokumenty
Z515930
Sigma-Aldrich
Aldrich® solvent storage flask
capacity 250 mL, joint: ST/NS 29/32
Synonyma:
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom
single-neck
joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
General description
With ground-glass joint and glass stopcock. Use septum Z565695 or Z565679 (see Septa Section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)