Key Documents
Z509221
Watch glasses
O.D. 65 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
concave
soda-lime glass
sterility
non-sterile
packaging
pkg of 12 ea
manufacturer/tradename
Sigma-Aldrich
O.D.
65 mm
General description
Concave, clear annealed soda lime glass with fire-polished edges.