Z507288
Michel-Miller Swagelok® adapter
NPTF 1/8 in., PTFE, Ace-Thred for #15
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
Doporučené produkty
material
PTFE
manufacturer/tradename
Ace Glass 584462
technique(s)
LPLC: suitable
NPTF
1/8 in.
fitting
Ace-Thred for #15
General description
Use to make Swagelok tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with #50 Ace-Thred fit Ace large columns.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)