Hlavní dokumenty
Z505137
Ace O-rings, FETFE®
I.D. 47.2 mm, wall size 3.5 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
packaging
pkg of 3 ea
manufacturer/tradename
Ace Glass 7855744
I.D.
47.2 mm
wall size
3.5 mm
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Zákazníci si také prohlíželi
Slide 1 of 1
1 of 1
Ace pressure tube bushing type, Back seal, capacity 200 mL, L × O.D. 20.3 cm × 50 mm
Z566276
Ace pressure tube