Z503797
Scienceware® trays
w/o faucet, inside dimensions L × W × H 30 cm × 41 cm × 7.6 cm
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Doporučené produkty
material
low-density polyethylene container
manufacturer/tradename
Bel-Art
General description
Molded LDPE can withstand temperatures up to 77°C (171°F). The PE faucet has a 12.7 mm bore.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)