Hlavní dokumenty
Z420565
Sigma-Aldrich
Coarse porous filter plate for Table-Top Buchner funnels
for Z420522
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
material
high-density polyethylene
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H91462-4091
disc diam.
36 in.
color
none
pore size
90—130 μm
General description
Porous filter plates are used with Table-Top Buchner funnels for quick filtration of large volumes. The plates is supported by a multiple ring grid; HDPE, 6.4mm (¼") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used.
Features and Benefits
  • Available in two porosities, medium and coarse
  • Medium is 45-90 microns
  • Coarse is 90-130 microns
  • Temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF)
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)