Z420395
Coarse porous filter plate for Table-Top Buchner funnels
for Z420360
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
Doporučené produkty
material
high-density polyethylene
description
For use with 18 in (45.7 cm) Buchner Table-Top Funnels
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H91462-4457
disc diam.
18 in.
filter diam.
18 in.
color
none
pore size
coarse
Související kategorie
General description
Porous filter plates are used with Table-Top Buchner funnels for quick filtration of large volumes. The plates is supported by a multiple ring grid; HDPE, 6.4mm (¼") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used.
Features and Benefits
- Available in two porosities, medium and coarse
- Medium is 45-90 microns
- Coarse is 90-130 microns
- Temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF)
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)