Key Documents
Z415529
Sigma-Aldrich
Aldrich® short-path distillation head with jacketed condenser
Joints: ST/NS 14/20 (2), thermometer joint: ST/NS 10/18
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
thermometer joint: ST/NS10/18
Joints: ST/NS14/20 (2)
condenser L
150 mm
immersion depth
50 mm