Key Documents
Z415502
Sigma-Aldrich
Aldrich® Claisen distillation head with Vigreux indents
Joints: ST/NS 14/20 (3), H 250 mm, thermometer joint: ST/NS 10/18
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
thermometer joint: ST/NS10/18
Joints: ST/NS14/20 (3)
H
250 mm
immersion depth
50 mm
General description
Vigreux indents run the entire length of the column and act as the packing for the column.