Key Documents
Z319988
Vapor duct for Büchi® model B-585 ovens
duct for 10, 20, and 40 mL flasks
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
feature
duct for 10, 20, and 40 mL flasks
manufacturer/tradename
Büchi 037073
General description
for use with B580 oven only
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG