Hlavní dokumenty
Z308811
Quickfit® conical flasks
capacity 250 mL, joint: ST/NS 19/26
Synonyma:
FE250/2
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
conical flask
flat bottom
joint
joint: ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
without cap
packaging
case of 5
manufacturer/tradename
Quickfit FE250/2
availability
not available in USA and Canada
capacity
250 mL
General description
With ground-glass socket for general use. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)