Z247227
Coors™ porcelain crucible cover
cap size C, I.D. 49
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
Doporučené produkty
cap size
C
packaging
pkg of 12 ea
manufacturer/tradename
Coorstek 66124
I.D.
49
General description
Coors™ porcelain crucible cover is glazed all over except for the rim.
Features and Benefits
- Ensures great resistance to thermal-shock
- Durable
- Rigid
- Sustained refractory qualities
Other Notes
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Legal Information
Coors is a trademark of Coorstek, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)