Z223565
Aldrich® septum-inlet adapter with stopcock and side-arm
Screw-thread, joint: ST/NS 14/20, glass stopcock
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Doporučené produkty
material
glass stopcock
joint
joint: ST/NS14/20
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)