Z220434
Sealing tube
capacity 15 mL, female joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Doporučené produkty
label
V
joint
female joint: ST/NS14/20
capacity
15 mL
Související kategorie
General description
For use with solid transfer vessel.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Zákazníci si také prohlíželi
Sortimentní položky
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.