Z180017
Michel-Miller column filter disc
Ace-Thred for #11
About This Item
UNSPSC Code:
12000000
material
paper
manufacturer/tradename
Ace Glass 581411
technique(s)
LPLC: suitable
fitting
Ace-Thred for #11
General description
Use at either end of column. Filter disc sizes 7 and 11 are made from specially pure, very uniform, highly-absorbent paper. Order adapter separately.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)