Key Documents
Z179965
Supelco
Stainless steel tubing adapter
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
manufacturer/tradename
Ace Glass 580912
technique(s)
LPLC: suitable
General description
Use with end-fitting adapter to connect tubing to M-M columns. Thread is 1/4-28 to 1.5mm O.D. extension.