Key Documents
Z177474
Sigma-Aldrich
Vakuwash®
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
technique(s)
UV/Vis spectroscopy: suitable
General description
For rinsing and drying glass cuvettes. Cell is held, open end down, against a rubber cushion which closes vacuum system. Includes 500 mL PP flask, Vikem® stopper, tubing, and Vakuwash funnel.
Legal Information
Vakuwash is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP