Key Documents
Z174998
ACE microscale round-bottom flask
size 10 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
single-neck
sterility
non-sterile
feature
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9592-10
Ace Glass 959210
size
10 mL