Key Documents
Z167479
Sigma-Aldrich
Round-bottom flask for Aldrich® student glass kit
for Z166898, capacity 25 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
short
single-neck
sterility
non-sterile
feature
closure type female Joint: ST/NS
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
25 mL
General description
14/20 female joint with outer screw thread