Key Documents
Z162930
Zipper-top poly bag
size 2 1/2 in. × 3 in.
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
material
clear low-density polyethylene
size
2 1/2 in. × 3 in.
thickness
4 mil
General description
Zipper-top poly bag is see-through with a double-track zipper. It meets FDA and USDA specifications for food contact. The bags also have a helpful hang hole at the top.