Hlavní dokumenty
Z148024
Sigma-Aldrich
Storage bottles with septum-inlet
capacity 500 mL, PTFE stopcock
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
PTFE stopcock
clear glass
round bottle
joint
joint ST/NS24/40 threaded
packaging
pack of 1 ea
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
General description
All septum inlets have 4 mm bore stopcocks requiring rubber septa, Z553913 or Z554030.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)