Z129593
Aldrich® micro sublimation apparatus
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Doporučené produkty
joint
joint: ST/NS24/40
General description
2 mm glass stopcock with O-ring. Use with recovery flask Z515574 or reaction tube Z516023.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)