Z116963
Stainless steel 304 syringe needle
gauge 12, L 6 in.
About This Item
UNSPSC Code:
41122002
eCl@ss:
32070500
NACRES:
NB.03
Doporučené produkty
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
6 in.
gauge
12
Související kategorie
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Standard Hub with 304 Stainless Steel Tubing
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)