Hlavní dokumenty
Z115398
Sigma-Aldrich
Spinbar® assortment
octagonal PTFE-coated, pack of 24 ea
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
octagonal PTFE-coated
packaging
pack of 24 ea
manufacturer/tradename
Bel-Art F37185-0001
General description
3 of each size in red, yellow, and blue: 1/2 × 1/8 in., 1/2 × 5/16 in., 5/8 × 5/16 in., 7/8 × 5/16 in., 1 × 5/16 in., 11/2 × 5/16 in., 2 × 5/16 in., 3 × 1/2 in.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)