Key Documents
Z112852
Sigma-Aldrich
PE bench-top base
size M
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
size
M
size
20 in. × 16 in.
thickness
1/2 in.
General description
BENCH-TOP BASE is not autoclavable and PP BENCH TRAY and rod are autoclavable.
Rigid. Keeps AtmosBag in place. Fits inside respective bag.