Z108294
Tempo-Shield heat-resistant open top shield
pack of 1 pair
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Doporučené produkty
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Sigma-Aldrich
General description
Optional, open-top shield (mitten) allows for additional short-term, high-temperature (up to 1,000 °F), exposure protection while permitting finger dexterity.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)