Hlavní dokumenty
WHA10443005
Whatman® glass vacuum filtration device
GV100 series, GV100/0/06 locking clamp with silicone seal, pack of 1
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
borosilicate glass funnel(s) (funnel)
stainless steel clamp(s) (clamp)
packaging
pack of 1
manufacturer/tradename
Whatman 10443005
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
General description
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Features and Benefits
  • Accessory to GV100 Glass Filter Funnel
Legal Information
Whatman is a registered trademark of Cytiva
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)