Key Documents
SYNC243445
Synthware Allihn reflux condenser
joint: ST/NS 34/45, jacket L 190 mm
Synonyma:
Synthware C243445, bulb condenser, Allihn condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
joint
joint: ST/NS34/45
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C243445
jacket L
190 mm
General description
Allihn reflux condenser with a water cooled inner drip joint at bottom.