Key Documents
SYNA851420
Synthware U-shaped drying tube
joint: ST/NS 14/20
Synonyma:
Synthware A851420, Drying tube 90° bent
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
colorless tube
glass tube
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A851420
General description
With joint. Cotton is usually used to contain drying agent.