Hlavní dokumenty
SYNA131024
Synthware Vigreux distillation connecting adapter
top joint: ST/NS 10/18, bottom joint: ST/NS 24/40, side joint: ST/NS 24/40, immersion depth 76 mm
Synonyma:
Synthware A131024, Distillation adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
joint
bottom joint: ST/NS24/40
side joint: ST/NS24/40
top joint: ST/NS10/18
packaging
pack of 1 ea
manufacturer/tradename
A131024
immersion depth
76 mm
Související kategorie
General description
Synthware Vigreux distillation connecting adapter with a top standard outer joint (10/18) to accommodate a 76mm immersion thermometer. With standardized 24/40 side and bottom inner joint (24/40), it easily fits various laboratory glassware.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)