Hlavní dokumenty
MT30207954
Mettler Toledo Seven2Go portable conductivity meter S3
Meter Only
About This Item
UNSPSC Code:
41115609
NACRES:
NB.61
description
connector LTW 7-pin (IP67)
pH/ion/ORP meters
feature
Meter Only
manufacturer/tradename
Mettler-Toledo 30207954
TDS measuring range
0.01 mg/L-300 g/L
conductivity measuring range
0.00-2022% (Ash)
0.01 μS/cm-500 mS/cm
range of application measuring range
0.00-100.0 Ω-cm (specific resistance)
salinity measuring range
0.00-42 (psu - practical salinity units)
temperature measuring range
-5-105 °C
battery power
4 x 1.5 V AA (or 1.3 V NiMH accumulators)
General description
Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
Simple Menu – Intuitive Operation:
  • Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions
T-Pad – Comfortable, Fast Navigation:
  • Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement
ISM® Ready – Reduced Chances for Error:
  • Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data
IP67 – Weatherproof and Robust Design:
  • Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
Features and Benefits
Includes: Meter electrode clip, instructions, quick guide, test certificate, declaration of conformity, batteries
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)