Key Documents
GF64417495
Brass
wire, Cu63%/Zn37%, 0.08 mm diameter, length 250 m, temper hard
Synonyma:
CU025120
About This Item
UNSPSC Code:
30102010
NACRES:
NA.23
form
wire
manufacturer/tradename
Goodfellow 644-174-95
L
250 m
diameter
0.08 mm