Key Documents
GF40614094
Polyethylene Naphthalate
(PEN), film, thickness 0.025 mm, size 300 × 300 mm, condition biaxially oriented
Synonyma:
ES361025
About This Item
UNSPSC Code:
12352100
NACRES:
NA.23
form
film
manufacturer/tradename
Goodfellow 406-140-94
size
300 × 300 mm
thickness
0.025 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c