Key Documents
GF00710750
Alumina
foam, thickness 7 mm, size 30 × 100 mm, porosity 0.86
Synonyma:
AL603820
About This Item
UNSPSC Code:
11181511
NACRES:
NA.23
manufacturer/tradename
Goodfellow 007-107-50
size
30 × 100 mm
thickness
7 mm