Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 630,00 Kč |
O této položce
NACRES:
NB.61
UNSPSC Code:
81141502
2 630,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliproduct line
SynLectro®
size
3 mm × 20 mm × 60 mm
General description
SynLectro® Copper Electrode is a 3x20x60 mm electrode designed for use with the SynLectro® Electrolysis Platform for synthetic electrochemistry.
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
Tato položka | |||
|---|---|---|---|
| description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm | description size 3 mm × 20 mm × 60 mm | description - |
| product line SynLectro® | product line SynLectro® | product line SynLectro® | product line SynLectro® |
| size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm | size 3 mm × 20 mm × 60 mm | size - |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?