DWKWCL1000-1
WHEATON® CELLine Multiuse Bioreactor flasks for suspension cell culture
polystyrene, flask capacity (1,000 mL), case of 1 ea
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
Doporučené produkty
material
molded polypropylene neck
polystyrene
screw top plastic cap
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 1 ea
manufacturer/tradename
WHEATON® WCL1000-1
flask capacity
(1,000 mL)
General description
CELLine Bioreactor Flask for high density suspension cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
- Disposable and ready-to-use
- High call density and high product concentration
- Bulk media chamber allows for longer run times without media refreshing and reduces operation time
- Decreases use of consumables
- Cost-efficient, space saving, and stackable
- No additional equipment required for operation
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)