Key Documents
DWK11-0056DB
Wheaton® Microliter Snap Caps With Septa
dark blue PTFE/silicone cap
About This Item
UNSPSC Code:
41100000
NACRES:
NB.11
material
dark blue PTFE/silicone cap
feature
cap pre-cut
cap starburst
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-0056DB
Application
Chromatography
Features and Benefits
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.