Hlavní dokumenty
CAD9918
Animal feeding needle, sterile, disposable, malleable 304 SS
size 18G, L × diam. 2 in. × 2.3 mm , ball
About This Item
UNSPSC Code:
42142609
NACRES:
NB.03
size
18G
packaging
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
2 in. × 2.3 mm , ball
General description
  • Designed to enable the researcher to shape the needle to specific needs
  • Needles come straight and are easily bent to any desired shape
  • Made of type 304 stainless steel tubing
  • Plastic Luer hubs
  • Silicone tips for smoothest possible insertion
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)