Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 790,00 Kč |
O této položce
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , glass
Sterility:
non-sterile
Wells:
1
1 790,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
colorless , glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 718605
technique(s)
cell culture: suitable
chamber size
0.1 mm , depth
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each.The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.The large square in the center is subdivided into 16 squares with 0.25 mm sides. Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm². The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.Contrary to the more advanced Improved Neubauer counting chambers, the counting area of each group square is limited by the outer one of the triple boundary lines.
Features and Benefits
- BLAUBRAND® quality
- Outstanding measurement precision
- 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Tato položka | |||
|---|---|---|---|
| description without clips, double ruled | description with clips, double ruled | description without clips, double ruled | description without clips, bright-line |
| manufacturer/tradename BRAND 718605 | manufacturer/tradename BRAND 718620 | manufacturer/tradename BRAND 717805 | manufacturer/tradename BRAND 717810 |
| material colorless , glass | material glass , colorless | material colorless , glass | material glass , colorless |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| wells 1 | wells 1 | wells 1 | wells 1 |
| feature without clips | feature with clips | feature without clips | feature without clips |
| chamber size 0.1 mm , depth | chamber size 0.1 mm , depth | chamber size 0.1 mm , depth | chamber size 0.1 mm , depth |
Still not finding the right product?
Explore all of our products under BRAND® counting chamber BLAUBRAND® Neubauer pattern
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?