Key Documents
BR28935
BRAND® Aluminum Pipet box
Holds 315-485 mm pipettes
About This Item
UNSPSC Code:
41123300
NACRES:
NB.13
material
aluminum
feature
Holds 315-485 mm pipettes
packaging
pack of 1 ea
manufacturer/tradename
BRAND 28935
General description
Pipette box of aluminum, inner silicone coating for storage and sterilisation of pipettes with different length.