Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 kit | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 26 800,00 Kč |
O této položce
NACRES:
NA.12
UNSPSC Code:
12352200
26 800,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliGeneral description
TLAM- Iδ1MεTγ-13CH3 kit has 13CH3 isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.
Application
For detection of long-range nOes or for the NMR study of large proteins
TLAM- Iδ1MεTγ-13CH3 kit is optimized with regio-specific labeling of isoleucine, methionine and threonine residues with 13CH3 isotopomer. This kit has been tested with protein isotopic labeling in E. coli. It is used in filtering inter-molecular NOEs in supramolecular assemblies and more precise structural information of homo-oligomeric protein structure.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.
Stable Isotopes Customer Service.
1 of 1
Tato položka | |||
|---|---|---|---|
| description - | description - | description - | description - |
| technique(s) bio NMR: suitable | technique(s) bio NMR: suitable | technique(s) bio NMR: suitable | technique(s) bio NMR: suitable |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage temp. −70°C | storage temp. −70°C | storage temp. −70°C | storage temp. −70°C |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?