D46904
N,N-Dibutylformamide
99%
Synonym(s):
N,N-Di-n-butylformamide, N-Formyldibutylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HCON(CH2CH2CH2CH3)2
CAS Number:
Molecular Weight:
157.25
Beilstein:
636100
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
<1 mmHg ( 20 °C)
Assay
99%
form
liquid
refractive index
n20/D 1.44 (lit.)
bp
120 °C/15 mmHg (lit.)
density
0.864 g/mL at 25 °C (lit.)
SMILES string
[H]C(=O)N(CCCC)CCCC
InChI
1S/C9H19NO/c1-3-5-7-10(9-11)8-6-4-2/h9H,3-8H2,1-2H3
InChI key
NZMAJUHVSZBJHL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hye Won Kim et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
Styrene can be formed by the microbial metabolism of bacteria and fungi. In our previous study, styrene was determined as a spoilage marker of Fuji apples decayed by Penicillium expansum, which is responsible for postharvest diseases. In the present study
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service