Key Documents
RF453011-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
F-45-30-11, non aerosol tight, to hold, 30 x 1.5/2.0 mL reaction vessels, lid
About This Item
UNSPSC Code:
41103909
feature
lid
to hold 30 x 1.5/2.0 mL reaction vessels
fixed angle rotor (microtubes)
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022637002 (NA)
Eppendorf® 5804715004 (EU)
parameter
20,800 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG